Learn More
Bioworld Agar, Noble, 500 g
Catalog No. 50255210
Encompass_Preferred
AgarUltra-high purityTrace amounts of ash and mineral contaminants removedGel Strength: 700−1200 g/cm2Working Concentration: 3−8 g/LGelling Temperature: 32.0−37.5 °CAsh Content: <1.5%This product is sold for laboratory use onlyAppearance: SolidHS Code : 130231000Melting Point: 80W22;95 °CPH Value: 5.8W22;7.0RTECS Number: AW7950000
Catalog No. 50-255-210 Supplier Bioworld Supplier No. 401000682
AgarUltra-high purityTrace amounts of ash and mineral contaminants removedGel Strength: 700−1200 g/cm2Working Concentration: 3−8 g/LGelling Temperature: 32.0−37.5 °CAsh Content: <1.5%This product is sold for laboratory use onlyAppearance: SolidHS Code : 130231000Melting Point: 80W22;95 °CPH Value: 5.8W22;7.0RTECS Number: AW7950000