Learn More
Research Products International Corp Agar, Noble, 1 KG
SDP
Catalog No. 501977047
Encompass_Preferred
Ultra-high purity Agar recommended for use in applications where maximum clarity and purity are essential. Thorough bleaching and washing procedures are utilized to remove trace amounts of ash and mineral contaminants.
Concentrations of 3-8 g/L obtains a semi-solid gel.
Catalog No. 50-197-7047 Supplier Research Products International Corp Supplier No. A204601000.0
Ultra-high purity Agar recommended for use in applications where maximum clarity and purity are essential. Thorough bleaching and washing procedures are utilized to remove trace amounts of ash and mineral contaminants.
Concentrations of 3-8 g/L obtains a semi-solid gel.