Learn More
Research Products International Corp Agar, Phyto 100 Grams, Specially Selected and Tested for Use in Plant Tissue Culture
SDP
Catalog No. 501978367
Encompass_Preferred
Specially selected and tested agar for use in plant tissue culture. High gel strength.
Concentration of 5.0 g/l obtains a solid gel strength: (1600 g/cm2).
Catalog No. 50-197-8367 Supplier Research Products International Corp Supplier No. A20300100.0
Specially selected and tested agar for use in plant tissue culture. High gel strength.
Concentration of 5.0 g/l obtains a solid gel strength: (1600 g/cm2).