Learn More
Research Products International Corp Agar, Phyto, High Gel Strength, 1 Kilogram
SDP
Catalog No. 50213129
Encompass_Preferred
Specially selected and tested agar for use in plant tissue culture. High gel strength.
Concentration of 5.0 g/l obtains a solid gel strength: (1600 g/cm2).
Catalog No. 50-213-129 Supplier Research Products International Corp Supplier No. A203001000.0
Specially selected and tested agar for use in plant tissue culture. High gel strength.
Concentration of 5.0 g/l obtains a solid gel strength: (1600 g/cm2).