Learn More
Alkali Scientific Agar, Phyto, High Gel Strength, 1 Kg
SDP
Catalog No. 5000660714
Encompass_Preferred
Phyto-grade agar with high gel strength is designed for plant culture applications requiring a firmer gel structure. This agar formulation is ideal for providing solid support during the growth of plant tissues, ensuring that the plant cultures remain stable and well-anchored in the medium. The 1kg package is ideal for larger-scale laboratory or industrial plant culture projects. With enhanced gel strength, this agar helps maintain consistency in plant growth environments, preventing cultures from losing their structure during incubation. It is widely used in plant biotechnology, breeding, and research to support the cultivation of a variety of plant species.
Catalog No. 50-006-60714 Supplier Alkali Scientific Supplier No. AGB003CP
Phyto-grade agar with high gel strength is designed for plant culture applications requiring a firmer gel structure. This agar formulation is ideal for providing solid support during the growth of plant tissues, ensuring that the plant cultures remain stable and well-anchored in the medium. The 1kg package is ideal for larger-scale laboratory or industrial plant culture projects. With enhanced gel strength, this agar helps maintain consistency in plant growth environments, preventing cultures from losing their structure during incubation. It is widely used in plant biotechnology, breeding, and research to support the cultivation of a variety of plant species.