Learn More
Bioworld Agar, Phytoagar, 1 kg
Catalog No. 50255213
Encompass_Preferred
AgarFree of extra salts that are often present in many agar preparations and affect plant growthGel Strength: 950 g/cm2Working Concentration: 5−7 g/LGelling Temperature: 38 °CMoisture: 10.92%Ash Content: 1.2%Swelling Index: 9.0 mLWater Absorption: 68.0 mLExtraneaous Matter: <0.3%HS Code : 130231000RTECS Number: AW7950000
Catalog No. 50-255-213 Supplier Bioworld Supplier No. 401000722
AgarFree of extra salts that are often present in many agar preparations and affect plant growthGel Strength: 950 g/cm2Working Concentration: 5−7 g/LGelling Temperature: 38 °CMoisture: 10.92%Ash Content: 1.2%Swelling Index: 9.0 mLWater Absorption: 68.0 mLExtraneaous Matter: <0.3%HS Code : 130231000RTECS Number: AW7950000