Learn More
Bioworld Agar, Phytoagar , 9002-18-0, 2.5kg
Supplier:  Bioworld 401000723
Encompass_Preferred
AgarFree of extra salts that are often present in many agar preparations and affect plant growth, Gel Strength: 950 g/cm2, Working Concentration: 5−7 g/L, Gelling Temperature: 38 °C, Moisture: 10.92%, Ash Content: 1.2%, Swelling Index: 9.0 mL, Water Absorption: 68.0 mL, Extraneaous Matter: <0.3%, HS Code : 130231000, RTECS Number: AW7950000, Molecular Formula : (C12H18O9)n, Solubility : H2O: 1.5% with heat, Insoluble in cold water and alcohol. Appearance Form : Powder, Appearance Color : White to light yellow. Melting Point : 85 - 95°C.
Catalog No. 50-255-214