Learn More
Research Products International Corp Agar, Plant, 1 Kilogram
SDP
Catalog No. 50213130
Encompass_Preferred
General purpose agar at an economical price. Concentration of 5.5 g/l obtains a solid gel strength: (1100 g/cm2).
Catalog No. 50-213-130 Supplier Research Products International Corp Supplier No. A204001000.0
General purpose agar at an economical price. Concentration of 5.5 g/l obtains a solid gel strength: (1100 g/cm2).