Learn More
Bioworld Agar, Plant Tissue Culture, 1 kg
Catalog No. 50255217
Encompass_Preferred
AgarGeneral purpose agar at an economical priceGel Strength: 1000 g/cm2Working Concentration: 7−9 g/LThis product is sold for laboratory research use only.HS Code : 130231000RTECS Number: AW7950000
Catalog No. 50-255-217 Supplier Bioworld Supplier No. 401000762
AgarGeneral purpose agar at an economical priceGel Strength: 1000 g/cm2Working Concentration: 7−9 g/LThis product is sold for laboratory research use only.HS Code : 130231000RTECS Number: AW7950000