Learn More
Biolog Inc BHI AGAR - Brain Heart Infusion Agar, 4 Plates/Pkg
SDP
Supplier:  Biolog Inc AS6426
Encompass
This item has a minimum qty of 4 per supplier requirements.
Brain Heart Infusion (BHI) Agar is a general purpose enriched non-selective media for the cultivation of a wide variety of microorganisms, including yeasts, molds, and bacteria. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.BHI Agar is an enriched non-selective media used for the isolation and cultivation of a wide variety of bacteria, including yeast and molds. The basic nutritive properties of this media are brain heart infusion from solids, meat peptone, and yeast extract. Dextrose provides a carbohydrate source for the fermentative microorganisms. Enrichment of the media with hemin and vitamin K1 is for the enhanced recovery of anaerobes.
Catalog No. NC1064252