Learn More
Teknova MacConkey Agar Plates with Levofloxacin-16. 100mm Plates, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. NC3668601
Encompass
MacConkey agar media aid in the isolation and differentiation of gram-negative bacterial isolates. It serves as an indicator, selective, and differential media, which determines the growth or presence of the gram-negative enteric bacilli from the different samples that are important for the sanitary purpose.
Catalog No. NC3668601 Supplier Teknova Supplier No. M1152
MacConkey agar media aid in the isolation and differentiation of gram-negative bacterial isolates. It serves as an indicator, selective, and differential media, which determines the growth or presence of the gram-negative enteric bacilli from the different samples that are important for the sanitary purpose.