Learn More
Electron Microscopy Sciences Agar, Powder 100 GR
SDP
Catalog No. 50980165
Encompass_Preferred
CAS #9002-18-0 (C₁₂H₁₈O9)x F.W. 3000-9000
Gelling Temperature (1.5% in H2O) about 35 degree C. Gel Strength (1.5% gel) 300g/cm2.
Catalog No. 50-980-165 Supplier Electron Microscopy Sciences Supplier No. 10200
CAS #9002-18-0 (C₁₂H₁₈O9)x F.W. 3000-9000
Gelling Temperature (1.5% in H2O) about 35 degree C. Gel Strength (1.5% gel) 300g/cm2.