Learn More
HIMEDIA Agar Powder, Bacteriological Grade, GRM026, Microbiology Culture Media Ingredients, Gelling Agent, From Red Seaweed, Bacteriological and Plant Tissue Culture, Powder, 500 g
Catalog No. NC4013639
Encompass_Preferred
Agar Powder, Bacteriological Grade is a natural gelling agent extracted from red seaweed under good manufacturing practice. It provides dependable gel strength for solidifying culture media and, with its higher mineral and metal content, is advantageous in formulations where ultra-high clarity and low ion content are not critical. It is recommended for general bacteriological culture media and for plant tissue culture media. An economical, reliable solidifying agent for the media kitchen. Store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013639 Supplier HIMEDIA Supplier No. GRM026500G
Agar Powder, Bacteriological Grade is a natural gelling agent extracted from red seaweed under good manufacturing practice. It provides dependable gel strength for solidifying culture media and, with its higher mineral and metal content, is advantageous in formulations where ultra-high clarity and low ion content are not critical. It is recommended for general bacteriological culture media and for plant tissue culture media. An economical, reliable solidifying agent for the media kitchen. Store in a cool dry place. Packaged as 500 g.