Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agar, Powder, FCC, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
9002-18-0
C14H24O9
336.34
GYYDPBCUIJTIBM-UHFFFAOYNA-N
2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
9002-18-0
C14H24O9
336.34
GYYDPBCUIJTIBM-UHFFFAOYNA-N
2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O

A1672, 9002-18-0

Spectrum™ Agar, Powder, FCC is a gel made from the cell walls of several species of red algae or seaweed. It appears as an odorless, tasteless, beige powder. It functions as a binder and thickener in cooking to make jellies, pudding, and custards. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Lead (Pb) 5 mg/kg
Packaging Fiber Drum
Quantity 25 kg