Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Agar, Simmons Citrate, 500 Grams
SDP

Catalog No. 501368593
Encompass_Preferred

Directions: Dissolve 24.2 g in 1L distilled water until evenly dispersed. Heat and stir until completely dissolved. Sterilize by autoclaving for 15 minutes. After autoclaving, place medium on a slant and allow to solidify.

Formulation per Liter:
Agar 15.0g
Ammonium Dihydrogen Phosphate 1.0g
Bromothymol Blue 0.08g
Magnesium Sulfate 0.2g
Potassium Phosphate Dibasic 1.0g
Sodium Chloride 5.0g
Sodium Citrate 2.0g
Final pH (25°C) 6.9±0.2
 

Catalog No. 50-136-8593 Supplier Research Products International Corp Supplier No. S29330500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Directions: Dissolve 24.2 g in 1L distilled water until evenly dispersed. Heat and stir until completely dissolved. Sterilize by autoclaving for 15 minutes. After autoclaving, place medium on a slant and allow to solidify.

Formulation per Liter:
Agar 15.0g
Ammonium Dihydrogen Phosphate 1.0g
Bromothymol Blue 0.08g
Magnesium Sulfate 0.2g
Potassium Phosphate Dibasic 1.0g
Sodium Chloride 5.0g
Sodium Citrate 2.0g
Final pH (25°C) 6.9±0.2
 

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.