Learn More
MilliporeSigma Agar Strips TCI-y, 40 units
Catalog No. NC1055131
Encompass
Agar Strips TCI-y are ready-to-use culture media for assessment of airborne microorganisms with RCS Air Samplers. Each individual agar strip is welded into a re-sealable sleeve and 100% visually controlled, ensuring maximum safety in critical environments.
Catalog No. NC1055131 Supplier MilliporeSigma Supplier No. 1442280040
Agar Strips TCI-y are ready-to-use culture media for assessment of airborne microorganisms with RCS Air Samplers. Each individual agar strip is welded into a re-sealable sleeve and 100% visually controlled, ensuring maximum safety in critical environments.