Learn More
MilliporeSigma Agar Strips YM Yeasts and molds (941196) HYCON 50/pk
Catalog No. 50727997
Encompass
Yeasts and molds (941196) HYCON Rose Bengal Agar with streptomycin; for determination of yeasts and molds. For use with RCS Microbial Air Samplers. Store at 2-25C.
Catalog No. 50-727-997 Supplier MilliporeSigma Supplier No. 1442420050
Yeasts and molds (941196) HYCON Rose Bengal Agar with streptomycin; for determination of yeasts and molds. For use with RCS Microbial Air Samplers. Store at 2-25C.