Learn More
Medchemexpress LLC Agaric acid (agaricinic acid) | 666-99-9 | MFCD00002665 | 99.5% | 416.55 g/mol | C22H40O7 | 10 MG
SDP
Catalog No. 5000289941
Encompass_Preferred
Agaric acid (agaricinic acid) is a naturally derived tricarboxylic small molecule used as a research reagent to investigate mitochondrial function and related metabolic processes. It has reported activity affecting mitochondrial permeability, citrate transport, fatty acid synthesis, and bacterial biofilm formation.
  • Orally active inhibitor of adenine nucleotide translocase.
  • Induces mitochondrial permeability transition and promotes calcium release.
  • Modulates citrate transport and fatty acid synthesis pathways.
  • Reported to inhibit bacterial biofilm formation.
  • Provided as a solid, high-purity reagent (99.45% reported) with recommended cold, light-protected storage.
Catalog No. 50-002-89941 Supplier Medchemexpress LLC Supplier No. HYN410410MG
Add to Cart
Add to Cart
Agaric acid (agaricinic acid) is a naturally derived tricarboxylic small molecule used as a research reagent to investigate mitochondrial function and related metabolic processes. It has reported activity affecting mitochondrial permeability, citrate transport, fatty acid synthesis, and bacterial biofilm formation.
  • Orally active inhibitor of adenine nucleotide translocase.
  • Induces mitochondrial permeability transition and promotes calcium release.
  • Modulates citrate transport and fatty acid synthesis pathways.
  • Reported to inhibit bacterial biofilm formation.
  • Provided as a solid, high-purity reagent (99.45% reported) with recommended cold, light-protected storage.