Learn More
Grainger Benchmark Scientific Agarose LE, Molecular Biology Grade, 100 g
Catalog No. 502749433
Encompass_Preferred
Agarose LE is a highly purified, organic solvent-free agarose engineered for high gel strength and integrity in molecular biology applications. It is suitable for analytical and preparative nucleic acid electrophoresis, protein electrophoresis, and various blotting protocols, supporting fragment sizes ranging from 150 bp to 25 kbp.
  • Low EEO (≤ 0.13 -mr) promotes increased electrophoretic mobility and shorter run times
  • Reduces band distortion caused by sulfate-rich negative ions
  • Highly transparent, forming a clear, colorless solution at 1 g:100 mL H2O
  • Free of nucleases, including RNase, DNase, and protease
  • Exceptional thermal stability and mechanical resistance for easy handling
  • Pore size adjustable via concentration modifications (0.5% to 2%)
Catalog No. 50-274-9433 Supplier Grainger Supplier No. 14N157
Agarose LE is a highly purified, organic solvent-free agarose engineered for high gel strength and integrity in molecular biology applications. It is suitable for analytical and preparative nucleic acid electrophoresis, protein electrophoresis, and various blotting protocols, supporting fragment sizes ranging from 150 bp to 25 kbp.
  • Low EEO (≤ 0.13 -mr) promotes increased electrophoretic mobility and shorter run times
  • Reduces band distortion caused by sulfate-rich negative ions
  • Highly transparent, forming a clear, colorless solution at 1 g:100 mL H2O
  • Free of nucleases, including RNase, DNase, and protease
  • Exceptional thermal stability and mechanical resistance for easy handling
  • Pore size adjustable via concentration modifications (0.5% to 2%)