Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Agarose

Catalog No. MP1AGAL0050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1AGAL0050 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1AGAL0050 Supplier MP Biomedicals™ Supplier No. 11AGAL0050
Add to Cart
Add to Cart

Agarose, Low Melting Point, Molecular Biology Grade, White Powder

  • Agarose low melting point is an intermediate melting and gelling point agarose which exhibits superior sieving characteristics.
  • It is recommended for analytical gels of DNA fragments under 1000 bp and is ideal for small DNA fragments down to 20 bp can be separated on 6 % gels.
  • It resolves fragments differing in size by only 2 bp.
  • Its use avoids the need for PAGE Southern and Northern transfers can be achieved efficiently from them at concentrations up to 2%.
  • DNA fragments can be purified with high efficiency from this agarose gels up to 4% by using either a purification kit or an electroelution system.
TRUSTED_SUSTAINABILITY

Specifications

CAS 9012-36-6
Color White
Form Powder
For Use With (Application) Nucleic Acidgel Electrophoresis & Blotting; Nucleic Acid Isolation & Purification; Nucleic Acid Electrophoresis; Nucleic Acid Sequencing
Melting Temperature < 67°C (1.5%gel)
Quantity 50 g
Storage Requirements Room Temperature
Product Type Agarose Gel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.