Learn More
Grainger Agarose 25g
Catalog No. 502749626
Encompass_Preferred
The Benchmark Scientific Agarose is a 25 gram container of molecular biology grade agarose powder. It has a gel point of 36-39 degrees Celsius and is ideal for DNA and RNA electrophoresis. This high purity agarose produces clear gels for excellent resolution. It can be used for techniques including PCR, restriction digests, and Southern and northern blotting. This agarose is sold in a plastic jar with a screw top lid.
Catalog No. 50-274-9626 Supplier Grainger Supplier No. 14N158
The Benchmark Scientific Agarose is a 25 gram container of molecular biology grade agarose powder. It has a gel point of 36-39 degrees Celsius and is ideal for DNA and RNA electrophoresis. This high purity agarose produces clear gels for excellent resolution. It can be used for techniques including PCR, restriction digests, and Southern and northern blotting. This agarose is sold in a plastic jar with a screw top lid.