Learn More
IBI SCIENTIFIC 3P 3:1 Super Sieve Agarose | 9012-36-6 | MFCD00081294 | 25 g
Catalog No. 50550456
Encompass_Preferred
IBI 3:1 Super Sieve is for analytical electrophoresis for high resolution gels. DNA 100-1500bp and PCR products <1000bp can be resolved. This agarose has a low viscosity so high concentration gels can be made with high resolving capacity.
Catalog No. 50-550-456 Supplier IBI SCIENTIFIC 3P Supplier No. IB70052
IBI 3:1 Super Sieve is for analytical electrophoresis for high resolution gels. DNA 100-1500bp and PCR products <1000bp can be resolved. This agarose has a low viscosity so high concentration gels can be made with high resolving capacity.