Learn More
Alkali Scientific ASI™ Agarose 3:1, 100g
SDP
Catalog No. 502389432
Encompass_Preferred
ASI™ Agarose 3:1 is blended high resolution agarose is designed for separation of fragments less than 1,000bp. Gels made with Agarose 3:1 are suitable for analytical and blotting applications. ASI™ Agarose 3:1 is manufactured under a “green” process that eliminates the use of organic solvents. They provide gels with enhanced resolution and clarity, low EEO and low background. Tested to be RNase, DNase, protease and endonuclease free. Supplied in 100g bottles.
Catalog No. 50-238-9432 Supplier Alkali Scientific Supplier No. A6603
ASI™ Agarose 3:1 is blended high resolution agarose is designed for separation of fragments less than 1,000bp. Gels made with Agarose 3:1 are suitable for analytical and blotting applications. ASI™ Agarose 3:1 is manufactured under a “green” process that eliminates the use of organic solvents. They provide gels with enhanced resolution and clarity, low EEO and low background. Tested to be RNase, DNase, protease and endonuclease free. Supplied in 100g bottles.