Learn More
Grainger Agarose 500g
Catalog No. 502749422
Encompass_Preferred
The Benchmark Scientific Agarose is a 500 gram container of molecular biology grade agarose powder. It has a gel point of 36-39 degrees Celsius and is used for DNA and RNA electrophoresis. The agarose powder can make 25 liters of 1% gel solution. It is high purity and RNase and DNase free. The agarose is ideal for separating nucleic acids and proteins. It produces clear gels for sharp band separation. This agarose is commonly used in research labs for gel electrophoresis procedures.
Catalog No. 50-274-9422 Supplier Grainger Supplier No. 14N156
The Benchmark Scientific Agarose is a 500 gram container of molecular biology grade agarose powder. It has a gel point of 36-39 degrees Celsius and is used for DNA and RNA electrophoresis. The agarose powder can make 25 liters of 1% gel solution. It is high purity and RNase and DNase free. The agarose is ideal for separating nucleic acids and proteins. It produces clear gels for sharp band separation. This agarose is commonly used in research labs for gel electrophoresis procedures.