Learn More
Medchemexpress LLC Agarose | 9012-36-6 | MFCD00081294 | Variable; ≈120,000 g/mol | C12H20O10(C12H18O9)n | 250 MG
SDP
Catalog No. 5000208482
Encompass_Preferred
Agarose low melting point is a laboratory-grade polysaccharide used for nucleic acid electrophoresis and for recovery of large or heat-labile DNA fragments. It melts at lower temperatures than standard agarose, facilitating gel extraction and enzymatic reactions with reduced thermal exposure.
  • low melting point suitable for gel extraction and enzymatic workflows.
  • form: powder that dissolves to form thermoreversible gels.
  • used for DNA and RNA electrophoresis and recovery of heat-labile samples.
  • stable as powder: store at 4°C to -20°C per manufacturer guidance.
  • supplied in small research pack sizes for molecular biology workflows.
Catalog No. 50-002-08482 Supplier Medchemexpress LLC Supplier No. 250MGHYW088070A
Add to Cart
Add to Cart
Agarose low melting point is a laboratory-grade polysaccharide used for nucleic acid electrophoresis and for recovery of large or heat-labile DNA fragments. It melts at lower temperatures than standard agarose, facilitating gel extraction and enzymatic reactions with reduced thermal exposure.
  • low melting point suitable for gel extraction and enzymatic workflows.
  • form: powder that dissolves to form thermoreversible gels.
  • used for DNA and RNA electrophoresis and recovery of heat-labile samples.
  • stable as powder: store at 4°C to -20°C per manufacturer guidance.
  • supplied in small research pack sizes for molecular biology workflows.