Learn More
Vector Laboratories Galanthus Nivalis (GNL GNA), Agarose Bound 5 ml
SDP
Supplier:  Vector Laboratories AL12435
Encompass
Galanthus nivalis lectin (GNL) preferentially binds to structures containing (a-1,3) mannose residues, such as many viral glycoproteins, most notably the gp120 coat protein of HIV. This agarose-bound GNL is prepared using affinity-purified lectin and heat stable, cross-linked 4% agarose beads. The attachment of the lectin through a hydrophilic spacer arm to the beads is carefully controlled to preserve the activity and minimize conformational changes to the bound lectins. The recommended inhibiting/eluting sugar is 100-200 mM a-methylmannoside.
Catalog No. NC1132296