Learn More
Bio Rad Laboratories AGAROSE GEL 1%+ 2X32
Catalog No. NC0999892
Encompass
1% Tae Wide Mini Readyagarose Precast Gel, 15.6 X 10 Cm, 2 X 32-Well, With Ethidium Bromide #1613054. 2 X 32-Well Comb Configuration, Precast Wide Mini Gel, Tae, 1.0% Plus Ethidium Bromide, Multichannel Pipet Compatible, 15.6 X 10 Cm (Tray Size). Choose Individually Packaged Wide Mini Readyagarose Precast Gels Made With High-Strength, High-Resolution Certified™ Agarose For All Your Molecular Biology Needs. The Ready-To-Load Gel Is In A Notched 15.6 X 10 Cm Uv-Transparent Tray That Holds The Gel In Place, And Can Be Loaded Using A Multichannel Pipet. Uv-Transparent Gel Tray; See And Record Results Without Handling The Gel. Trays Have Fluorescent Well Numbers And A Printed Ruler For Easy Reference. Lock Tabs To Prevent Movement During Loading And Running. 2-Sided Tray For Less Distortion Than A 4-Sided Tray Giving Consistently Straight Lanes. Gels Fit Securely In Bio-Rad'S Wide Mini-Sub® Cell Gt Cells, Or Run In Any Sub Cell At Least 15.6 Cm Wide.
Catalog No. NC0999892 Supplier Bio Rad Laboratories Supplier No. 1613054
1% Tae Wide Mini Readyagarose Precast Gel, 15.6 X 10 Cm, 2 X 32-Well, With Ethidium Bromide #1613054. 2 X 32-Well Comb Configuration, Precast Wide Mini Gel, Tae, 1.0% Plus Ethidium Bromide, Multichannel Pipet Compatible, 15.6 X 10 Cm (Tray Size). Choose Individually Packaged Wide Mini Readyagarose Precast Gels Made With High-Strength, High-Resolution Certified™ Agarose For All Your Molecular Biology Needs. The Ready-To-Load Gel Is In A Notched 15.6 X 10 Cm Uv-Transparent Tray That Holds The Gel In Place, And Can Be Loaded Using A Multichannel Pipet. Uv-Transparent Gel Tray; See And Record Results Without Handling The Gel. Trays Have Fluorescent Well Numbers And A Printed Ruler For Easy Reference. Lock Tabs To Prevent Movement During Loading And Running. 2-Sided Tray For Less Distortion Than A 4-Sided Tray Giving Consistently Straight Lanes. Gels Fit Securely In Bio-Rad'S Wide Mini-Sub® Cell Gt Cells, Or Run In Any Sub Cell At Least 15.6 Cm Wide.