Learn More
Research Products International Corp Agarose Gel Loading Dye 6X Solution Glycerol Based, 50 Milliliters
SDP
Catalog No. 50995774
Encompass_Preferred
6X Concentrated gel loading dye for molecular biology applications.
Formulation:
Bromophenol Blue: 0.25%
Xylene Cyanol FF: 0.25%
Glycerol: 40% in sterile water
Catalog No. 50-995-774 Supplier Research Products International Corp Supplier No. A2150050.0
6X Concentrated gel loading dye for molecular biology applications.
Formulation:
Bromophenol Blue: 0.25%
Xylene Cyanol FF: 0.25%
Glycerol: 40% in sterile water