Learn More
Bioworld Agarose Gel Loading Dye, (6X), Alkaline, 25 mL
Catalog No. 50196766
Encompass_Preferred
Agarose Gel Loading DyeGel loading dye for molecular biologyAlkalineSpecifications:6X concentration300 mM NaOH6 mM EDTA0.25% Bromocresol Green0.25% Xylene cyanol FF18% Ficoll in water
Catalog No. 50-196-766 Supplier Bioworld Supplier No. 105700011
Agarose Gel Loading DyeGel loading dye for molecular biologyAlkalineSpecifications:6X concentration300 mM NaOH6 mM EDTA0.25% Bromocresol Green0.25% Xylene cyanol FF18% Ficoll in water