Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Agarose Gel Loading Dye (6X, Glycerol-Based)

Catalog No. AAJ63869AD
Change view
Click to view available options
Quantity:
25 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63869AD 50 mL
AAJ63869AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63869AD Supplier Thermo Scientific Chemicals Supplier No. J63869AD
Add to Cart

missing translation for 'validMainframeMsg'

Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.

Solubility
Fully miscible in water.

Notes
0.25% bromophenol blue, 0.25% xylene cyanol FF and 40% glycerol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Agarose gel loading dye
Physical Form Liquid
Name Note glycerol based
Concentration 6X
Quantity 50 mL
MDL Number MFCD00675058
Solubility Information Fully miscible in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.