Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Agarose Gel Loading Dye (6X, Glycerol-Based, no Xylene Cyanol FF)

Catalog No. AAJ60933AD
Change view
Click to view available options
Quantity:
25 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60933AD 50 mL
AAJ60933AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60933AD Supplier Thermo Scientific Chemicals Supplier No. J60933AD
Add to Cart

missing translation for 'validMainframeMsg'

6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.

Solubility
Fully misicible in water.

Notes
No Xylene cyanol FF. Contains 0.25% bromophenol blue, 30% glycerol.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Agarose gel loading dye
Physical Form Liquid
Name Note glycerol based, Xylene Cyanol FF free
Concentration 6X
Quantity 50 mL
Solubility Information Fully misicible in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.