Learn More
Bioworld Agarose Gel Loading Dye, (6X), Alkaline, 50 mL
Catalog No. 50196767
Encompass_Preferred
Agarose Gel Loading DyeGel loading dye for molecular biologyAlkalineSpecifications:6X concentration300 mM NaOH6 mM EDTA0.25% Bromocresol Green0.25% Xylene cyanol FF18% Ficoll in water
Catalog No. 50-196-767 Supplier Bioworld Supplier No. 105700012
Agarose Gel Loading DyeGel loading dye for molecular biologyAlkalineSpecifications:6X concentration300 mM NaOH6 mM EDTA0.25% Bromocresol Green0.25% Xylene cyanol FF18% Ficoll in water