Learn More
Bioworld Agarose Gel Loading Dye (6X) Glycerol with BB, 25 mL
Catalog No. 50196772
Encompass_Preferred
Gel loading dye for use in agarose gel electrophoresis for molecular biology Prepared in sterile 3D water., Store at RT.Technical Information:6X concentrationBromophenol blue (0.25%)Xylene cyanol FF (0.25%)Glycerol in water (30%)Storage: 2-8C
Catalog No. 50-196-772 Supplier Bioworld Supplier No. 105700041
Gel loading dye for use in agarose gel electrophoresis for molecular biology Prepared in sterile 3D water., Store at RT.Technical Information:6X concentrationBromophenol blue (0.25%)Xylene cyanol FF (0.25%)Glycerol in water (30%)Storage: 2-8C