Learn More
Bioworld Agarose Gel Loading Dye (6X) with Sucrose, 25 mL
Catalog No. 50196770
Encompass_Preferred
Agarose Gel Loading Dye, 6X, Sucrose Gel loading dye for molecular biology Technical Information:6X concentrationContains Bromophenol Blue (0.25%)Xylene cyanol FF (0.25%)Sucrose (40% w/v)Storage: 2-8C
Catalog No. 50-196-770 Supplier Bioworld Supplier No. 105700031
Agarose Gel Loading Dye, 6X, Sucrose Gel loading dye for molecular biology Technical Information:6X concentrationContains Bromophenol Blue (0.25%)Xylene cyanol FF (0.25%)Sucrose (40% w/v)Storage: 2-8C