Learn More
Bioworld Agarose Gel Loading Dye (6X) with Sucrose, 50 mL
Catalog No. 50196771
Encompass_Preferred
Agarose Gel Loading Dye, 6X, Sucrose Gel loading dye for molecular biology Technical Information:6X concentrationContains Bromophenol Blue (0.25%)Xylene cyanol FF (0.25%)Sucrose (40% w/v)Storage: 2-8C
Catalog No. 50-196-771 Supplier Bioworld Supplier No. 105700032
Add to Cart
Add to Cart
Agarose Gel Loading Dye, 6X, Sucrose Gel loading dye for molecular biology Technical Information:6X concentrationContains Bromophenol Blue (0.25%)Xylene cyanol FF (0.25%)Sucrose (40% w/v)Storage: 2-8C