Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Agarose Gel Reagents: TBE Buffer
SCI_ED

Catalog No. S20652 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S20652 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S20652 Supplier Innovating Science™ Supplier No. IS5206
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5X TBE buffer concentrate

Chemical Identifiers

CAS 77-86-1 , 7732-18-5
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Concentration 10X
For Use With (Application) Gel Electrophoresis and Blotting
pH 8.3
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Percent Purity 5x
Grade Reagent
Chemical Name or Material TBE Buffer 5XConcentrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.