Learn More
Embi Tec Runone Casting Tray | 4 mini | 2 Landscape
SDP
Catalog No. NC1606364
Encompass
Agarose Gel Tray Set for RunOne Casting System that includes 4 mini (5.4x5.9cm) and 2 landscape (10.9x5.9mg) printed polycarbonate trays.
Catalog No. NC1606364 Supplier Embi Tec Supplier No. EP1005
Agarose Gel Tray Set for RunOne Casting System that includes 4 mini (5.4x5.9cm) and 2 landscape (10.9x5.9mg) printed polycarbonate trays.