Learn More
Alkali Scientific Agarose HR, High Resolution, 500 Grams
SDP
Supplier:  Alkali Scientific SEH001CP
Encompass_Preferred
Agarose HR (High Resolution) is a specialized agarose used in DNA gel electrophoresis for sharp band separation. With a high-resolution profile, it offers superior performance in separating small DNA fragments, providing clearer and more distinct bands than standard agarose. This 500g package is ideal for labs that frequently perform high-resolution separation for genetic analysis, PCR applications, or sequencing. Agarose HR is designed to maintain structural integrity during the gel preparation process, ensuring precise results in applications such as DNA analysis, restriction digestion analysis, and other molecular biology applications requiring high-fidelity gel electrophoresis.
Catalog No. 50-006-60971