Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Agarose I 500 mg Tablets
SDP

Catalog No. 5024469
Encompass_Preferred

CAS #9012-36-6 (CHOCHO)n
Melting point: 87-89C

Agarose I is standard melting/gelling agarose, suitable for routine nucleic acid analytical/preservative applications. Agarose I has a low EEO for shorter electrophoresis runs without compromising resolution. Excellent for blotting technique and general manipulations.
pH: 5.7 7.7 (3%, Water) @ 25C

Catalog No. 50-244-69 Supplier Electron Microscopy Sciences Supplier No. 10206
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #9012-36-6 (CHOCHO)n
Melting point: 87-89C

Agarose I is standard melting/gelling agarose, suitable for routine nucleic acid analytical/preservative applications. Agarose I has a low EEO for shorter electrophoresis runs without compromising resolution. Excellent for blotting technique and general manipulations.
pH: 5.7 7.7 (3%, Water) @ 25C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.