Learn More
Bioworld AGAROSE I BIOTECH GRADE
Supplier:  Bioworld 401001043
Encompass_Preferred
Agarose 1, Biotech Grade bioPlus Agarose I is formulated specifically for analysis of nucleic acids, using standard electrophoretic techniques. Specifications: Biotechnology Grade; Gel strength: 1200g/cm2; Gelling point: 33-39C; Mel...
Catalog No. 50-255-227