Learn More
Bioworld AGAROSE I BIOTECH GRADE
Supplier: Bioworld 401001044
Agarose 1, Biotech Grade bioPlus Agarose I is formulated specifically for analysis of nucleic acids, using standard electrophoretic techniques. Specifications: Biotechnology Grade; Gel strength: 1200g/cm2; Gelling point: 33-39C; Mel...