Learn More
Bioworld Agarose I Tablets, 50 g
Supplier:  Bioworld 401001441
Encompass_Preferred
Save time and labor preparing agarose gels for electrophoresis of nucleic acids. Just place the required numbers of tablets into a standard buffer, then mix and heat until completely dissolved.* Gel Strength, 1.5% >1200b/cm2* Gelling Range: 36-39°C* Melting Temp: 87-89°C* EEO: <0.10* Sulfate: <0.15* DNase, RNase None detectedUsually ships within 24 hours.This product is sold for laboratory use only. Appearance: SolidHS Code : 130239000
Catalog No. 50-255-247