Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agarose (Isoelectric Focusing of Proteins/IEF Grade), Fisher BioReagents

Catalog No. BP16325 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP163-25 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP163-25

missing translation for 'validMainframeMsg'

Highly purified agarose for separating proteins by isoelectric focusing.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Powder/Solid
CAS 9012-36-6
Color Cream
Quantity 25 g
DNase DNase free
Electroendosmosis (EEO) Not Detected
Gelation Temperature 34° to 45°C
Gel Strength 500g/cm2min.
Packaging Poly Bottle
Molecular Formula C12H18O9
Formula Weight 306.12
Grade EIF
Chemical Name or Material Agarose, Isoelectric Focusing of Proteins
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.