Learn More
Gold Biotechnology Inc Agarose LE (Molecular Biology Grade) 1 kg
SDP
Catalog No. 501532727
Encompass_Preferred
Agarose LE (Low Electroendosmosis) is Molecular Biology Grade and is specifically designed for the superior separation of nucleic acids, with maximally crisp band resolution, and is also ideal for cloning and cloning related experiments.
Catalog No. 50-153-2727 Supplier Gold Biotechnology Inc Supplier No. A2011000
Agarose LE (Low Electroendosmosis) is Molecular Biology Grade and is specifically designed for the superior separation of nucleic acids, with maximally crisp band resolution, and is also ideal for cloning and cloning related experiments.