Learn More
Gold Biotechnology Inc Agarose LE
SDP
Catalog No. 501532728
Encompass_Preferred
Agarose LE (Low Electroendosmosis) is Molecular Biology Grade and is specifically designed for the superior separation of nucleic acids with maximally crisp band resolution and is also ideal for cloning and cloning related experiments
Catalog No. 50-153-2728 Supplier Gold Biotechnology Inc Supplier No. A201500
Agarose LE (Low Electroendosmosis) is Molecular Biology Grade and is specifically designed for the superior separation of nucleic acids with maximally crisp band resolution and is also ideal for cloning and cloning related experiments