Learn More
Sigma Aldrich Fine Chemicals Biosciences Agarose LE low electroendo
Catalog No. 502906213
Encompass
Agarose is obtained from seaweed and contains repeated agarobiose units. During agarose gel formation the polymers aggregate to form a network with varying pore sizes. This property is used in agarose gel electrophoresis to separate DNA.
Catalog No. 50-290-6213 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11685660001
Agarose is obtained from seaweed and contains repeated agarobiose units. During agarose gel formation the polymers aggregate to form a network with varying pore sizes. This property is used in agarose gel electrophoresis to separate DNA.