Learn More
Sigma Aldrich Fine Chemicals Biosciences Agarose LE low electroendo
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11685678001
Encompass
Agarose is obtained from seaweed and contains repeated agarobiose units. During agarose gel formation the polymers aggregate to form a network with varying pore sizes. This property is used in agarose gel electrophoresis to separate DNA.
Catalog No. 50-290-2913