Learn More
Electron Microscopy Sciences Agarose LE Powder 100 GR
SDP
Catalog No. 501927937
Encompass_Preferred
EMS Agarose LE is a highly purified agarose refined using an advanced process that excludes the use of organic solvents, yielding a cleaner end-product with a significantly reduced environmental impact. It is widely used for nucleic acid electrophoresis (analytical and preparative), protein electrophoresis (including radial immunodiffusion) and various blotting protocols.
It's low EEO (≤ 0.13 -mr) promotes increased electrophoretic mobility, yielding improved resolution and shorter run times. This also allows macromolecules and larger particles (subcellular fragments, viruses, etc.) to migrate more freely through the gel matrix. The consistently low EEO also provides a reduction in band distortion (caused by counterflow) that can result from the presence of excessive sulfate-rich negative ions.
Catalog No. 50-192-7937 Supplier Electron Microscopy Sciences Supplier No. 8704602
EMS Agarose LE is a highly purified agarose refined using an advanced process that excludes the use of organic solvents, yielding a cleaner end-product with a significantly reduced environmental impact. It is widely used for nucleic acid electrophoresis (analytical and preparative), protein electrophoresis (including radial immunodiffusion) and various blotting protocols.
It's low EEO (≤ 0.13 -mr) promotes increased electrophoretic mobility, yielding improved resolution and shorter run times. This also allows macromolecules and larger particles (subcellular fragments, viruses, etc.) to migrate more freely through the gel matrix. The consistently low EEO also provides a reduction in band distortion (caused by counterflow) that can result from the presence of excessive sulfate-rich negative ions.