Learn More
Biotium Agarose LE, Ultra-Pure Molecular Biology Grade, 500 g
Supplier:  Biotium 41028500G
Encompass_Preferred
Ultra-pure LE Agarose can be used for all routine electrophoresis applications. It has low electroendosmosis (EEO) which confers high electrophoretic mobility. This agarose has excellent performance for analytical or preparative nucleic acid electrophoresis and blotting. Ultra-pure LE Agarose has high strength and shows low background after staining with common nucleic acid dyes. Specifications: Moisture: ≤ 10%; Ash: ≤ 0.4%; EEO: 0.05-0.13; Sulfate: ≤0.1%; Clarity (1.5% gel): ≤ 3 NTU; Gel strength (1% gel): ≥1200 g/cm²; Gel strength (1.5% gel): ≥ 2500 g/cm²; Gelling temperature (1.5% agarose): 36 ± 1.5 ⁰C; Melting temperature (1.5% gel): 88 ± 1.5 ⁰C; DNAse/RNase activity: None detected; DNA resolution ≥ 1000bp: finely resolved; Gel background: very low; Performance in GelRed precast DNA staining: verified; Performance in GelGreen precast dsDNA staining: verified; Performance in DNAzure visible blue dsDNA staining: verified.
Catalog No. 50-196-4677