Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Agarose LM
GREENER_CHOICE

Catalog No. 14223083 Shop All Axygen Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-223-083 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-223-083 Supplier Axygen™ Supplier No. AGRLM100
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Axygen™ Low melting agarose allows for the recovery of undamaged nucleic acids below their denaturation temperature.

The low gelling temperature ensures that the agarose will be in a liquid state at a temperature range where In-Gel manipulations can be performed without prior extraction of the DNA from the gel slice. Low Melting (LM) Agarose is derivatized by organic synthesis which generates methoxylate groups from the basic agarose structure. Agarose LM is also optimized for preparative electrophoresis and isolation of DNA fragments.
  • Highest gelling/melting temperatures and gel strength
  • Electrophoresis of DNA fragments ≥1000bp
  • Suitable for tissue and cell culture and viral plaque assays
  • Ideal for preparative electrophoresis
TRUSTED_SUSTAINABILITY

Specifications

Dnase Free Yes
For Use With (Application) Tissue and cell culture and viral plaque assays
Material Agarose
Melting Temperature ≥76°C
Quantity 100 g
RNase Free Yes
Storage Requirements 15° to 25°C
Product Type LM Agarose Gel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.